Invitrogen
Manufacturer Code:PA534046
Catalog # PIPA534046
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 14 amino acid peptide from 2nd extracellular domain of human OR10R2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Olfactory receptor 10R2; Olfactory receptor OR1-8; olfactory receptor, family 10, subfamily R, member 2; OR10R2 Olfactory receptor OR1-8 OR1-8 OR10R2Q Olfactory receptor 10R2
Gene Aliases: OR1-8; OR10R2; OR10R2Q
UniProt ID: (Human) Q8NGX6
Entrez Gene ID: (Human) 343406
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.