Invitrogen
Manufacturer Code:PA5102269
Catalog # PIPA5102269
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human OR10R2 corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Olfactory receptor 10R2; Olfactory receptor 10R2 olfactory receptor family 10 subfamily R member 2 Olfactory receptor OR1-8 olfactory receptor family 10 subfamily R member 2 OR10R2 OR10R2Q OR1-8; Olfactory receptor OR1-8; olfactory receptor, family 10, subfamily R, member 2
Gene Aliases: OR1-8; OR10R2; OR10R2Q
UniProt ID: (Human) Q8NGX6
Entrez Gene ID: (Human) 343406
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.