Abnova
Manufacturer Code:PAB2593
Catalog # 89-127-560
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to N-terminus of human OPRS1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of OPRS1.
For Research Use Only
Protein Aliases: Aging-associated gene 8 protein; FLJ25585; MGC3851; OPRS1; SIG-1R; Sigma 1-type opioid receptor; Sigma non-opioid intracellular receptor 1; SR-BP; SR-BP1; SR31747 binding protein 1; SR31747-binding protein
Gene Aliases: AAG8; ALS16; DSMA2; hSigmaR1; OPRS1; SIG-1R; sigma1R; SIGMAR1; SR-BP; SR-BP1; SRBP
UniProt ID: (Human) Q99720
Entrez Gene ID: (Human) 10280
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.