Invitrogen
Manufacturer Code:PA526138
Catalog # PIPA526138
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 161-187 amino acids from the central region of human OPRM1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: LMOR MOR MOR1 OPRM M-OR-1 MOP hMOP mu opiate receptor mu opioid receptor hMOR-1a mu opioid receptor splice variant hMOR-1S mu opioid receptor splice variant hMOR-1Y2 mu opioid receptor splice variant hMOR-1Y3 mu opioid receptor splice variant hMOR-1Z mu opioid receptor spli; M-OR-1; MOP; MOP receptor; Mu opiate receptor; Mu opioid receptor; mu opioid receptor hMOR-1a; mu opioid receptor splice variant mMOR-1Z; Mu-type opioid receptor; opioid receptor, mu 1
Gene Aliases: LMOR; M-OR-1; MOP; MOP-R; MOR; MOR-1; MOR-1O; MOR1; muOR; OPRM; OPRM1
UniProt ID: (Human) P35372, (Mouse) P42866
Entrez Gene ID: (Human) 4988, (Mouse) 18390
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.