Invitrogen
Manufacturer Code:PA14561
Catalog # PIPA14561
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide comprising residues N(386) H Q L E N L E A E T A P L P(400) of human mouse and rat MOR-1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: M-OR-1; MOP; MOP receptor; MOR-1; Mu opiate receptor; Mu opioid receptor; mu opioid receptor hMOR-1a; mu opioid receptor splice variant mMOR-1Z; mu opioid receptor splice variant rMOR-1S; mu opioid receptor splice variant rMOR-1Z; Mu-type opioid receptor; MUOR1; Opioid receptor B; opioid receptor, mu 1; OPRM1 Opioid Receptor Mu-1
Gene Aliases: LMOR; M-OR-1; MOP; MOP-R; MOR; MOR-1; MOR-1O; MOR1; MORA; muOR; OPRM; OPRM1; Oprrm1; Ror-b
UniProt ID: (Human) P35372, (Rat) P33535, (Mouse) P42866
Entrez Gene ID: (Human) 4988, (Rat) 25601, (Mouse) 18390
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.