Abnova
Manufacturer Code:PAB12257
Catalog # 89-115-681
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to residues surrounding S375 of mouse Oprm1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Oprm1.
For Research Use Only
Protein Aliases: M-OR-1; MOP receptor; MOP-R; mor; MOR-1; MOR-1O; mu opioid receptor splice variant mMOR-1Z; Mu-type opioid receptor; muOR; Oprm
Gene Aliases: M-OR-1; MOP-R; Mor; MOR-1; MOR-1O; muOR; Oprm; Oprm1
UniProt ID: (Mouse) P42866
Entrez Gene ID: (Mouse) 18390
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.