Invitrogen
Manufacturer Code:PA533861
Catalog # PIPA533861
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic 18 amino acid peptide from C-terminus of human Kappa Opioid Receptor. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K-OR-1; kappa opioid receptor; Kappa-type opioid receptor; KOR-1; Opiate receptor, kappa-1; opioid receptor, kappa 1; OPRK1 Kappa-type opioid receptor Opiate receptor kappa-1 K-OR-1 KOR Opioid receptor kappa 1 OPRK Ork1 Kappa opioid receptor KOR-1 Opioid receptor kappa
Gene Aliases: K-OR-1; KOR; KOR-1; OPRK; OPRK1; Ror-d
UniProt ID: (Human) P41145, (Rat) P34975
Entrez Gene ID: (Human) 4986, (Rat) 29335
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.