Invitrogen
Manufacturer Code:PA124535
Catalog # PIPA124535
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide - KLH conjugated. Sequence identity/similarity to Mouse: 88/100&percnt. Rat: 88/100&percnt. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: K-OR-1; Kappa-type opioid receptor; KOR-1; MSL-1; opioid receptor, kappa 2
Gene Aliases: K-OR-1; KOR; KOR-1; MSL-1; Oprk1; Oprk2; R21; Ror-d
UniProt ID: (Mouse) P33534, (Rat) P34975
Entrez Gene ID: (Mouse) 18387, (Rat) 29335
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.