Abnova
Manufacturer Code:PAB18980
Catalog # 89-120-533
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids at C-terminus of human OPRK1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Goat polyclonal antibody raised against synthetic peptide of OPRK1.
For Research Use Only
Protein Aliases: K-OR-1; kappa opioid receptor; Kappa-type opioid receptor; KOR; Opiate receptor, kappa-1; opioid receptor, kappa 1; OPRK
Gene Aliases: K-OR-1; KOR; KOR-1; OPRK; OPRK1
UniProt ID: (Human) P41145
Entrez Gene ID: (Human) 4986
Molecular Function:
G-protein coupled receptor
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.