Abnova
Manufacturer Code:PAB18160
Catalog # 89-119-733
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (PFA fixed) (IHC (PFX)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to mouse Oprk1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Oprk1.
For Research Use Only
Protein Aliases: K-OR-1; Kappa-type opioid receptor; KOR; KOR-1; MGC151172; MSL-1; opioid receptor, kappa 2; Oprk2; R21
Gene Aliases: K-OR-1; KOR; KOR-1; MSL-1; Oprk1; Oprk2; R21
UniProt ID: (Mouse) P33534
Entrez Gene ID: (Mouse) 18387
Molecular Function: G-protein coupled receptor receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.