Invitrogen
Manufacturer Code:MA527154
Catalog # PIMA527154
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI2F5 |
Immunogen | Full length human recombit protein of OPA3 produced in E.coli |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Optic atrophy 3 (autosomal recessive with chorea and spastic paraplegia) FLJ22187 FLJ25932 MGA3 MGC75494; Optic atrophy 3 (Iraqi-Jewish 'optic atrophy plus'); Optic atrophy 3 protein
Gene Aliases: MGA3; OPA3
UniProt ID: (Human) Q9H6K4
Entrez Gene ID: (Human) 80207
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.