Novus Biologicals
Manufacturer Code:NBP171656
Catalog # NBP171656
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1E8-1D9 |
Immunogen | Human OPA1 [Swiss-Prot# O60313]. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: Dynamin-like 120 kDa protein, form S1; Dynamin-like 120 kDa protein, mitochondrial; dynamin-like guanosine triphosphatase; FLJ12460 KIAA0567dynamin-like 120 kDa protein mitochondrial MGM1 mitochondrial dynamin-like GTPase NPGlargeG NTGmitochondrial dynamin-like 120 kDa protein optic atrophy 1 (autosomal dominant) Optic atrophy protein 1; mitochondrial dynamin-like GTPase; optic atrophy 1 (autosomal dominant); Optic atrophy protein 1
Gene Aliases: BERHS; KIAA0567; largeG; MGM1; MTDPS14; NPG; NTG; OPA1
UniProt ID: (Human) O60313
Entrez Gene ID: (Human) 4976
Molecular Function: G-protein cytoskeletal protein enzyme modulator hydrolase microtubule family cytoskeletal protein small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.