Abnova
Manufacturer Code:H00004976M05
Catalog # 89-123-261
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1C10 |
Immunogen | OPA1 (NP_056375 851 a.a. ~ 960 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant OPA1.
For Research Use Only
Protein Aliases: Dynamin-like 120 kDa protein, form S1; Dynamin-like 120 kDa protein, mitochondrial; dynamin-like guanosine triphosphatase; FLJ12460; KIAA0567; largeG; MGM1; mitochondrial dynamin-like GTPase; NPG; NTG; optic atrophy 1 (autosomal dominant); Optic atrophy protein 1
Gene Aliases: BERHS; KIAA0567; largeG; MGM1; MTDPS14; NPG; NTG; OPA1
UniProt ID: (Human) O60313
Entrez Gene ID: (Human) 4976
Molecular Function: G-protein cytoskeletal protein enzyme modulator hydrolase microtubule family cytoskeletal protein small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.