R&D Systems
Manufacturer Code:MAB9506SP
Catalog # MAB9506SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | 1284B |
Immunogen | A synthetic peptide made to an internal region within amino acid residues 500-600 of human OPA1 Accession # O60313 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rabbit MonoclonalAntibody
For Research Use Only
Protein Aliases: BERHS EC 3.6.5.5 FLJ12460 KIAA0567dynamin-like 120 kDa protein mitochondrial LargeG lilr3 MGM1 mitochondrial dynamin-like GTPase MTDPS14 NPG NPGlargeG NTG NTGmitochondrial dynamin-like 120 kDa protein optic atrophy 1 (autosomal dominant) Optic atrophy protein 1; Dynamin-like 120 kDa protein, form S1; Dynamin-like 120 kDa protein, mitochondrial; dynamin-like guanosine triphosphatase; mitochondrial dynamin-like GTPase; optic atrophy 1 (autosomal dominant); Optic atrophy protein 1
Gene Aliases: BERHS; KIAA0567; largeG; MGM1; MTDPS14; NPG; NTG; OPA1
UniProt ID: (Human) O60313
Entrez Gene ID: (Human) 4976
Molecular Function: G-protein cytoskeletal protein enzyme modulator hydrolase microtubule family cytoskeletal protein small GTPase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.