Novus Biologicals
Manufacturer Code:NBP231658
Catalog # NBP231658
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against a recombinant protein corresponding to amino acids: MLTRGEQHLSRGLGTPPAAMMSHLNGLHHPGHTQSHGPVLAPSRERPPSSSSGSQVATSGQLEE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hepatocyte nuclear factor 6-beta; HNF-6-beta; OC-2; One cut domain family member 2; one cut domain family member 2 OC2 OC-2 one cut homeobox 2; one cut domain, family member 2; One cut homeobox 2; onecut 2; ONECUT-2 homeodomain transcription factor; Transcription factor ONECUT-2
Gene Aliases: HNF6B; OC-2; OC2; ONECUT2
UniProt ID: (Human) O95948
Entrez Gene ID: (Human) 9480
Molecular Function:
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.