Invitrogen
Manufacturer Code:PA5114396
Catalog # PIPA5114396
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence of human ONECUT2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: C730009D12 Hepatocyte nuclear factor 6-beta HNF6B HNF-6B HNF-6-beta Oc2 OC-2 One cut domain family member 2 one cut domain family member 2 one cut homeobox 2 onecut 2 ONECUT2 ONECUT-2 homeodomain transcription factor RGD1564677 transcription factor ONECUT2 Transcription factor ONECUT-2; Hepatocyte nuclear factor 6-beta; HNF-6-beta; OC-2; One cut domain family member 2; one cut domain, family member 2; One cut homeobox 2; onecut 2; ONECUT-2 homeodomain transcription factor; Transcription factor ONECUT-2; transcription factor ONECUT2
Gene Aliases: C730009D12; HNF6B; OC-2; OC2; ONECUT2
UniProt ID: (Human) O95948, (Mouse) Q6XBJ3
Entrez Gene ID: (Human) 9480, (Mouse) 225631
Molecular Function:
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.