Proteintech
Manufacturer Code:219161AP150UL
Catalog # 50-173-3638
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | ONECUT2 Fusion Protein Ag16162 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Hepatocyte nuclear factor 6-beta; HNF 6 beta HNF6B OC 2 OC2 One cut domain family member 2 one cut homeobox 2 ONECUT2 Transcription factor ONECUT 2; HNF-6-beta; OC-2; One cut domain family member 2; one cut domain, family member 2; One cut homeobox 2; onecut 2; ONECUT-2 homeodomain transcription factor; Transcription factor ONECUT-2; transcription factor ONECUT2
Gene Aliases: C730009D12; HNF6B; OC-2; OC2; ONECUT2
UniProt ID: (Human) O95948, (Mouse) Q6XBJ3
Entrez Gene ID: (Human) 9480, (Mouse) 225631
Molecular Function:
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.