Invitrogen
Manufacturer Code:PA5113975
Catalog # PIPA5113975
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the middle region of human OMA1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2010001O09Rik DAB1 Metalloendopeptidase OMA1 mitochondrial metalloprotease related protein 1 Metalloprotease-related protein 1 MPRP1 MPRP-1 OMA1 OMA1 homolog zinc metallopeptidase OMA1 homolog zinc metallopeptidase (S. cerevisiae) OMA1 zinc metallopeptidase OMA1 zinc metallopeptidase homolog overlapping activity with M-AAA protease Overlapping with the m-AAA protease 1 homolog peptidase RGD1304821 YKR087C zinc metallopeptidase homolog zinc metallopeptidase OMA1 ZMPOMA1; Metalloendopeptidase OMA1, mitochondrial; metalloprotease related protein 1; Metalloprotease-related protein 1; MPRP-1; OMA1 homolog, zinc metallopeptidase; OMA1 zinc metallopeptidase homolog; overlapping activity with M-AAA protease; Overlapping with the m-AAA protease 1 homolog; zinc metallopeptidase OMA1
Gene Aliases: 2010001O09Rik; DAB1; MPRP-1; MPRP1; OMA1; peptidase; YKR087C; ZMPOMA1
UniProt ID: (Human) Q96E52
Entrez Gene ID: (Human) 115209
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.