Novus Biologicals
Manufacturer Code:NBP156970100UL
Catalog # N156970100U
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to OMA1 (OMA1 homolog zinc metallopeptidase (S. cerevisiae)) The peptide sequence was selected from the middle region of OMA1. Peptide sequence WAICPRDSLALLCQWIQSKLQEYMFNRPYSRKLEAEADKIGLLLAAKACA. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
Protein Aliases: 2010001O09Rik DAB1 EC 3.4.24.- metalloprotease related protein 1 Metalloprotease-related protein 1 MPRP1 MPRP-1mitochondrial OMA1 homolog zinc metallopeptidase (S. cerevisiae) overlapping activity with M-AAA protease YKR087C ZMPOMA1; Metalloendopeptidase OMA1, mitochondrial; metalloprotease related protein 1; Metalloprotease-related protein 1; MPRP-1; OMA1 homolog, zinc metallopeptidase; OMA1 zinc metallopeptidase homolog; overlapping activity with M-AAA protease; Overlapping with the m-AAA protease 1 homolog; zinc metallopeptidase OMA1
Gene Aliases: 2010001O09Rik; DAB1; MPRP-1; MPRP1; OMA1; peptidase; YKR087C; ZMPOMA1
UniProt ID: (Human) Q96E52
Entrez Gene ID: (Human) 115209
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.