Invitrogen
Manufacturer Code:PA568726
Catalog # PIPA568726
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | synthetic peptide directed towards the middle region of human OLIG3 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: basic helix-loop-helix domain-containing class B protein 7; bHLHb7; BHLHB7 BHLHB7 Bhlhb7 bHLHb7 BHLHE20 bHLHe20oligo3 Class B basic helix-loop-helix protein 7 Class E basic helix-loop-helix protein 20 Oligo3 oligodendrocyte transcription factor 3; bHLHe20; Class B basic helix-loop-helix protein 7; Class E basic helix-loop-helix protein 20; Oligo3; oligodendrocyte transcription factor 2; Oligodendrocyte transcription factor 3; Oligodendrocyte-specific bHLH transcription factor 3
Gene Aliases: BHLHB7; BHLHE20; OLIG3
UniProt ID: (Human) Q7RTU3, (Mouse) Q6PFG8
Entrez Gene ID: (Dog) 484005, (Rabbit) 100349761, (Human) 167826, (Horse) 100146586, (Mouse) 94222, (Rat) 293012, (Guinea pig) 100722252
Molecular Function: basic helix-loop-helix transcription factor nuclease nucleic acid binding transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.