Novus Biologicals
Manufacturer Code:NBP224535
Catalog # NBP224535
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to amino acids between 400 and 450 of human OLFM4 was used as immunogen for this antibody. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antiapoptotic protein GW112; Antiapoptotic protein GW112 bA209J19.1 GC1 G-CSF-stimulated clone 1 protein GW112hOLfD hGC-1 KIAA4294 olfactomedin 4 olfactomedin-4 OlfD OLM4 UNQ362; G-CSF-stimulated clone 1 protein; hGC-1; hOLfD; Olfactomedin-4; OLM4
Gene Aliases: bA209J19.1; GC1; GW112; hGC-1; hOLfD; OlfD; OLFM4; OLM4; UNQ362; UNQ362/PRO698
UniProt ID: (Human) Q6UX06
Entrez Gene ID: (Human) 10562
Molecular Function:
receptor
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.