Invitrogen
Manufacturer Code:PA5110059
Catalog # PIPA5110059
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant protein of human OLFM3. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: B230206G02Rik Noe3 NOELIN3 Noelin-3 Olfactomedin-3 OLFM3 Optimedin UNQ1924/PRO4399; Noelin-3; olfactomedin related ER localized protein 3; Olfactomedin-3; Optimedin
Gene Aliases: B230206G02Rik; NOE3; NOELIN3; OLFM3; OPTIMEDIN; UNQ1924/PRO4399
UniProt ID: (Human) Q96PB7, (Mouse) P63056, (Rat) P63057
Entrez Gene ID: (Human) 118427, (Mouse) 229759, (Rat) 252920
Molecular Function:
receptor
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.