Abnova
Manufacturer Code:H00118427A01
Catalog # 89-107-574
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Polyclonal |
Type | Antibody |
Immunogen | 108 a.a. &bsim 206 a.a) partial recombinant protein with GST tag.,OLFM3 (NP_477518 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse polyclonal antibody raised against a partial recombinant OLFM3.
For Research Use Only
Protein Aliases: NOE3; Noelin-3; NOELIN3; NOELIN3_V1; NOELIN3_V2; NOELIN3_V3; NOELIN3_V4; NOELIN3_V5; NOELIN3_V6; olfactomedin related ER localized protein 3; Olfactomedin-3; OPTIMEDIN
Gene Aliases: NOE3; NOELIN3; OLFM3; OPTIMEDIN; UNQ1924/PRO4399
UniProt ID: (Human) Q96PB7
Entrez Gene ID: (Human) 118427
Molecular Function:
receptor
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.