Invitrogen
Manufacturer Code:PA548087
Catalog # PIPA548087
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Sheep |
Class | Polyclonal |
Type | Antibody |
Immunogen | CHO-derived recombit human Olfactomedin-2/Noelin-2 Gln21-Pro454 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Sheep Polyclonal Antibody
For Research Use Only
Protein Aliases: neuronal olfactomedin related ER localized protein 2; NOE2 NOELIN2 NOELIN2&lowbarV1 OlfC neuronal olfactomedin related ER localized protein 2 noelin 2 noelin-2 olfactomedin-2; Noelin-2; Olfactomedin-2
Gene Aliases: NOE2; NOELIN2; NOELIN2_V1; OlfC; OLFM2
UniProt ID: (Human) O95897
Entrez Gene ID: (Human) 93145
Molecular Function: receptor structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.