Abnova
Manufacturer Code:MAB11613
Catalog # 89-109-765
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S96-7 |
Immunogen | A synthetic peptide corresponding to amino acids 28-48 of human OLFM1. |
Conjugate | Biotin |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C.<br />Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against synthetic peptide of OLFM1.
For Research Use Only
Protein Aliases: AMY; neuroblastoma protein; Neuronal olfactomedin-related ER localized protein; NOE1; Noelin; NOELIN1; OlfA; olfactomedin related ER localized protein; Olfactomedin-1; pancortin 1
Gene Aliases: AMY; NOE1; NOEL1; NOELIN1; OlfA; OLFM1
UniProt ID: (Human) Q99784
Entrez Gene ID: (Human) 10439
Molecular Function:
receptor
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.