Invitrogen
Manufacturer Code:PA528842
Catalog # PIPA528842
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide corresponding to a region within amino acids 421 and 485 of Human Noelin |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AMY NOE1 NOELIN1 OlfA neuroblastoma protein neuronal olfactomedin-related ER localized protein noelin olfactomedin related ER localized protein olfactomedin-1 pancortin 1; neuroblastoma protein; Neuronal olfactomedin-related ER localized protein; Noelin; Noelin 1; Noelin 2; olfactomedin related ER localized protein; Olfactomedin-1; Pancortin; pancortin 1; Pancortin 1-4
Gene Aliases: AMY; AW742568; NOE1; Noel; NOEL1; NOELIN1; OlfA; OLFM1; Pancortin; Pancortin3
UniProt ID: (Human) Q99784, (Mouse) O88998
Entrez Gene ID: (Human) 10439, (Mouse) 56177
Molecular Function:
receptor
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.