Abnova
Manufacturer Code:PAB2559
Catalog # 89-127-205
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide (conjugated with KLH) corresponding to amino acids 441-471 at C-terminus of rat Olfm1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of Olfm1.
For Research Use Only
Protein Aliases: 1B426B; D2Sut1e; Neuronal olfactomedin-related ER localized protein; Noe1; Noelin; olfactomedin related ER localized protein; Olfactomedin-1; Pancortin
Gene Aliases: D2Sut1e; Noe1; Noel; Noel1; Olfm1
UniProt ID: (Rat) Q62609
Entrez Gene ID: (Rat) 93667
Molecular Function:
receptor
structural protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.