Invitrogen
Manufacturer Code:PA558179
Catalog # PIPA558179
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human OLAH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Augmented in rheumatoid arthritis 1; AURA1; Oleoyl-ACP hydrolase; oleoyl-acyl-carrier-protein hydrolase; S-acyl fatty acid synthase thioesterase, medium chain; TE2; Thioesterase 2; thioesterase domain containing 1; Thioesterase domain-containing protein 1; Thioesterase II
Gene Aliases: AURA1; OLAH; SAST; THEDC1
UniProt ID: (Human) Q9NV23
Entrez Gene ID: (Human) 55301
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.