Invitrogen
Manufacturer Code:PA595319
Catalog # PIPA595319
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to a sequence at the C-terminus of human OGT identical to the related mouse and rat sequences. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: O linked N-acetylglucosamine transferase; O-GlcNAc transferase p110 subunit; O-GlcNAc transferase subunit p110; O-linked N-acetylglucosamine (GlcNAc) transferase (UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase); O-linked N-acetylglucosamine transferase 110 kDa subunit; OGT; UDP-N-acetylglucosamine--peptide N-acetylglucosaminyltransferase 110 kDa subunit; UDP-N-acetylglucosamine--peptide N-acetylglucosaminyltransferase 110 kDa subunit 2.4.1.255 O-GlcNAc transferase subunit p110 O-linked N-acetylglucosamine transferase 110 kDa subunit OGT OGT; UDP-N-acetylglucosamine:polypeptide-N-acetylglucosaminyl transferase; uridinediphospho-N-acetylglucosamine:polypeptide beta-N-acetylglucosaminyl transferase
Gene Aliases: 1110038P24Rik; 4831420N21Rik; AI115525; HINCUT-1; HRNT1; O-GLCNAC; OGT; Ogtl
UniProt ID: (Human) O15294, (Mouse) Q91Y38, (Rat) P56558
Entrez Gene ID: (Human) 8473, (Mouse) 108155, (Rat) 26295
Molecular Function: glycosyltransferase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.