Proteintech
Manufacturer Code:663821IG150UL
Catalog # 50-173-6811
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2A5F11 |
Immunogen | OGN Fusion Protein Ag3484 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: corneal keratan sulfate proteoglycan; Mimecan; Mimecan OG OGN OIF osteoglycin Osteoinductive factor SLRR3A; mimecan proteoglycan; OIF; Osteoglycin; osteoglycin/mimecan; Osteoinductive factor
Gene Aliases: OG; OGN; OIF; SLRR3A
UniProt ID: (Human) P20774
Entrez Gene ID: (Pig) 106509723, (Human) 4969
Molecular Function:
extracellular matrix protein
receptor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.