Invitrogen
Manufacturer Code:MA535616
Catalog # PIMA535616
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Monoclonal |
Type | Antibody |
Clone | ARC1233 |
Immunogen | A synthesized peptide derived from human OGG1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Monoclonal Antibody
For Research Use Only
Protein Aliases: 8-hydroxyguanine DNA glycosylase; 8-oxoguanine DNA glycosylase; 8-oxoguanine DNA glycosylase AP lyase DNA-(apurinic or apyrimidinic site) lyase HMMH HOGG1 Mmh MUTM N-glycosylase/DNA lyase OGG1 Ogh1; AP lyase; DNA-(apurinic or apyrimidinic site) lyase; DNA-apurinic or apyrimidinic site lyase; N-glycosylase/DNA lyase; OGG1 type 1f
Gene Aliases: HMMH; HOGG1; MMH; MUTM; OGG1; OGH1
UniProt ID: (Human) O15527, (Mouse) O08760
Entrez Gene ID: (Human) 4968, (Mouse) 18294
Molecular Function: DNA binding protein DNA glycosylase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.