Novus Biologicals
Manufacturer Code:NBP252724F
Catalog # NBP252724F
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 2B4 |
Immunogen | Partial recombinant protein made to an internal sequence of the human OGG1 protein (between amino acids 100-300) [Uniprot: O15527] |
Conjugate | FITC |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: 8-hydroxyguanine DNA glycosylase; 8-hydroxyguanine DNA glycosylase 8-oxoguanine DNA glycosylase AP lyase DNA-apurinic or apyrimidinic site lyase HOGG1 MMH MUTMOGH1HMMH N-glycosylase/DNA lyase; 8-oxoguanine DNA glycosylase; AP lyase; DNA-(apurinic or apyrimidinic site) lyase; DNA-apurinic or apyrimidinic site lyase; N-glycosylase/DNA lyase; OGG1 type 1f
Gene Aliases: HMMH; HOGG1; MMH; MUTM; OGG1; OGH1
UniProt ID: (Human) O15527
Entrez Gene ID: (Human) 4968
Molecular Function:
DNA binding protein
DNA glycosylase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.