Novus Biologicals
Manufacturer Code:NB100106SS
Catalog # NB100106SS
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A peptide derived from human Ogg1 (within amino acids 1-100). [UniProt# O15527] |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 8-hydroxyguanine DNA glycosylase; 8-hydroxyguanine DNA glycosylase 8-oxoguanine DNA glycosylase AP lyase DNA-apurinic or apyrimidinic site lyase HOGG1 MMH MUTMOGH1HMMH N-glycosylase/DNA lyase; 8-oxoguanine DNA glycosylase; AP lyase; DNA-(apurinic or apyrimidinic site) lyase; DNA-apurinic or apyrimidinic site lyase; N-glycosylase/DNA lyase; OGG1 type 1f
Gene Aliases: HMMH; HOGG1; MMH; MUTM; OGG1; OGH1
UniProt ID: (Human) O15527
Entrez Gene ID: (Human) 4968
Molecular Function:
DNA binding protein
DNA glycosylase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.