Proteintech
Manufacturer Code:151251AP150UL
Catalog # 50-173-0001
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | OGG1 Fusion Protein Ag7204 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 8 oxoguanine DNA glycosylase AP lyase HMMH HOGG1 MMH MUTM OGG1 OGH1; 8-hydroxyguanine DNA glycosylase; 8-oxoguanine DNA glycosylase; 8-oxoguanine DNA-glycosylase 1; 8-oxoguanine-DNA-glycosylase; AP lyase; DNA-(apurinic or apyrimidinic site) lyase; DNA-apurinic or apyrimidinic site lyase; N-glycosylase/DNA lyase; OGG1 type 1f
Gene Aliases: HMMH; HOGG1; MMH; MUTM; OGG1; OGH1
UniProt ID: (Human) O15527, (Mouse) O08760, (Rat) O70249
Entrez Gene ID: (Human) 4968, (Mouse) 18294, (Rat) 81528
Molecular Function:
DNA binding protein
DNA glycosylase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.