Invitrogen
Manufacturer Code:PA521102
Catalog # PIPA521102
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A 17 amino acid peptide near the center of human OGFOD2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-oxoglutarate and iron-dependent oxygenase domain containing 2; 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 2
Gene Aliases: 1300006G11Rik; 5730405M13Rik; AU041636; AW552395; OGFOD2; RGD1310594
UniProt ID: (Human) Q6N063, (Mouse) Q9CQ04
Entrez Gene ID: (Human) 79676, (Mouse) 66627, (Rat) 288657
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.