Invitrogen
Manufacturer Code:PA5113038
Catalog # PIPA5113038
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 2 protein (1-186AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1300006G11Rik 2-oxoglutarate and iron dependent oxygenase domain containing 2 2-oxoglutarate and iron-dependent oxygenase domain containing 2 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 2 5730405M13Rik AU041636 AW552395 Ogfod2 RGD1310594; 2-oxoglutarate and iron-dependent oxygenase domain containing 2; 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 2
Gene Aliases: 1300006G11Rik; 5730405M13Rik; AU041636; AW552395; OGFOD2
UniProt ID: (Human) Q6N063, (Mouse) Q9CQ04
Entrez Gene ID: (Human) 79676, (Mouse) 66627
Molecular Function:
RNA binding protein
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.