Novus Biologicals
Manufacturer Code:NBP276391
Catalog # NBP276391
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Immunogen sequence between amino acids 1 and 50 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 2-8°C/ 1 year from date of receipt |
Rabbit Polyclonal Antibody
Protein Aliases: 2-oxoglutarate and iron-dependent oxygenase domain containing 1; 2-oxoglutarate and iron-dependent oxygenase domain containing 12-oxoglutarate and iron-dependent oxygenase domain-containing protein 1 EC 1.14.11 FLJ10826 KIAA1612TPA1 termination and polyadenylation 1 homolog Termination and polyadenylation 1 homolog TPA1EC 1.14.11.-; 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 1; Prolyl 3-hydroxylase OGFOD1; Termination and polyadenylation 1 homolog; TPA1, termination and polyadenylation 1, homolog; uS12 prolyl 3-hydroxylase
Gene Aliases: KIAA1612; OGFOD1; TPA1
UniProt ID: (Human) Q8N543
Entrez Gene ID: (Human) 55239
Molecular Function:
oxidoreductase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.