Novus Biologicals
Manufacturer Code:NBP297921DL594
Catalog # NB049478
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Produced in rabbits immunized with E. coli-derived Human OGFOD1 fragment. |
Conjugate | DyLight 594 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-oxoglutarate and iron-dependent oxygenase domain containing 1; 2-oxoglutarate and iron-dependent oxygenase domain containing 12-oxoglutarate and iron-dependent oxygenase domain-containing protein 1 EC 1.14.11 FLJ10826 KIAA1612TPA1 termination and polyadenylation 1 homolog Termination and polyadenylation 1 homolog TPA1EC 1.14.11.-; 2-oxoglutarate and iron-dependent oxygenase domain-containing protein 1; Prolyl 3-hydroxylase OGFOD1; Termination and polyadenylation 1 homolog; TPA1, termination and polyadenylation 1, homolog; uS12 prolyl 3-hydroxylase
Gene Aliases: KIAA1612; OGFOD1; TPA1
UniProt ID: (Human) Q8N543
Entrez Gene ID: (Human) 55239
Molecular Function: oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.