Proteintech
Manufacturer Code:171101AP150UL
Catalog # 50-173-1387
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | OGDHL Fusion Protein Ag10794 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-oxoglutarate dehydrogenase complex component E1-like; 2-oxoglutarate dehydrogenase-like, mitochondrial; Alpha-ketoglutarate dehydrogenase-like; KIAA1290 OGDC E1 like OGDHL; OGDC-E1-like
Gene Aliases: KIAA1290; OGDHL
UniProt ID: (Human) Q9ULD0, (Rat) D3ZQD3
Entrez Gene ID: (Human) 55753, (Mouse) 239017, (Rat) 290566
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.