Proteintech
Manufacturer Code:152121AP150UL
Catalog # 50-173-0069
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | OGDH Fusion Protein Ag7368 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 2-oxoglutarate dehydrogenase complex component E1; 2-oxoglutarate dehydrogenase E1 component, mitochondrial; 2-oxoglutarate dehydrogenase, mitochondrial; AKGDH E1k OGDC OGDC E1 OGDH; Alpha-ketoglutarate dehydrogenase; OGDC-E1; oxoglutarate (alpha-ketoglutarate) dehydrogenase (lipoamide); oxoglutarate decarboxylase; oxoglutarate dehydrogenase (lipoamide); oxoglutarate dehydrogenase (succinyl-transferring); testicular tissue protein Li 131
Gene Aliases: 2210403E04Rik; 2210412K19Rik; AA409584; AKGDH; d1401; E1k; Kiaa4192; mKIAA4192; OGDC; OGDH; Ogdhl
UniProt ID: (Human) Q02218, (Mouse) Q60597, (Rat) Q5XI78
Entrez Gene ID: (Human) 4967, (Mouse) 18293, (Rat) 360975
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.