Invitrogen
Manufacturer Code:PA5112634
Catalog # PIPA5112634
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant Human Oral-facial-digital syndrome 1 protein (598-771AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 71-7A BOS_26373 CXorf5 DXGgc7e JBTS10 OFD1 OFD1 centriole and centriolar satellite protein ofd1 protein OFD1 centriole and centriolar satellite protein oral-facial-digital syndrome 1 oral-facial-digital syndrome 1 gene homolog oral-facial-digital syndrome 1 gene homolog (human) Oral-facial-digital syndrome 1 protein Oral-facial-digital syndrome 1 protein homolog ORF2 Protein 71-7A putative oral-facial-digital syndrome 1 protein RGD1562231 RP23 SGBS2 wu:fa07a10 wu:fd17e11 zgc:92562; Oral-facial-digital syndrome 1 protein; Oral-facial-digital syndrome 1 protein homolog; Protein 71-7A
Gene Aliases: 71-7A; CXorf5; DXGgc7e; JBTS10; OFD1; ORF2; RP23; SGBS2
UniProt ID: (Human) O75665, (Mouse) Q80Z25
Entrez Gene ID: (Human) 8481, (Mouse) 237222
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.