Novus Biologicals
Manufacturer Code:NBP185417
Catalog # NBP185417
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:HKRGMKGDTVNVRRSVRVKTKNPPHCLEITPPSSEKLVSVMRLSDLSTEDDDSGHCKMNRYDKKIDSLMNAVGCLKSEVKMQKGERQMAKRFLEERKEELEEVAHELAETEHENTVLRHNIERMKEEKDFTILQKKHLQQEKE |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: cancer/testis antigen 134; Cenexin; cenexin 1; Cenexin FLJ44866 MGC111096 MGC9034 ODF2/1 ODF2/2 ODF8484-kD outer dense fiber of sperm tails 2 outer dense fiber protein 2 sperm tail structural protein; Outer dense fiber of sperm tails protein 2; outer dense fiber of sperm tails, 84-kD; Outer dense fiber protein 2; sperm tail structural protein; testis tissue sperm-binding protein Li 51e
Gene Aliases: CT134; ODF2; ODF2/1; ODF2/2; ODF84
UniProt ID: (Human) Q5BJF6
Entrez Gene ID: (Human) 4957
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.