Abnova
Manufacturer Code:PAB10187
Catalog # 89-114-685
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthetic peptide corresponding to C-terminus of human ODF2. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C. For long term storage store at -20°C.<br />Aliquot to avoid repeated freezing and thawing. |
Rabbit polyclonal antibody raised against synthetic peptide of ODF2.
For Research Use Only
Protein Aliases: cancer/testis antigen 134; Cenexin; cenexin 1; FLJ44866; MGC111096; MGC9034; ODF2/1; ODF2/2; ODF84; Outer dense fiber of sperm tails protein 2; outer dense fiber of sperm tails, 84-kD; Outer dense fiber protein 2; sperm tail structural protein; testis tissue sperm-binding protein Li 51e
Gene Aliases: CT134; ODF2; ODF2/1; ODF2/2; ODF84
UniProt ID: (Human) Q5BJF6
Entrez Gene ID: (Human) 4957
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.