Invitrogen
Manufacturer Code:MA125811
Catalog # PIMA125811
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 3F293 |
Immunogen | Synthetic hexadecapeptide corresponding to residues K(435) P D E K Y Y S S S I W G P T C(360) of rat ODC. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: ODC; Ornithine decarboxylase; Ornitine decarboxylase
Gene Aliases: ODC; ODC1
UniProt ID: (Human) P11926, (Rat) P09057, (Mouse) P00860
Entrez Gene ID: (Human) 4953, (Rat) 24609, (Mouse) 18263
Molecular Function:
decarboxylase
lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.