Novus Biologicals
Manufacturer Code:NBP294155100UL
Catalog # NB022867
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 42-141 of human SLC22A4 (NP_003050.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°:C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Ergothioneine transporter; Ergothioneine transporter ET transporter ETT MGC34546 MGC40524 OCTN1integral membrane transport protein Organic cation/carnitine transporter 1 solute carrier family 22 (organic cation/ergothioneine transporter) member 4 solute carrier family 22 member 4 UT2H; ET transporter; integral membrane transport protein; Organic cation/carnitine transporter 1; solute carrier family 22 (organic cation/ergothioneine transporter), member 4; solute carrier family 22 (organic cation/zwitterion transporter), member 4; Solute carrier family 22 member 4
Gene Aliases: ETT; OCTN1; SLC22A4; UT2H
UniProt ID: (Human) Q9H015
Entrez Gene ID: (Human) 6583
Molecular Function:
carbohydrate transporter
cation transporter
transfer/carrier protein
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.