Invitrogen
Manufacturer Code:PA537882
Catalog # PIPA537882
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-HEDGHEAQLEPSP from the internal region of POU3F1 (aa192-204) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: Oct-6; Octamer-binding protein 6; Octamer-binding transcription factor 6; OTF-6; POU domain transcription factor SCIP; POU domain, class 3, transcription factor 1; POU3F1 POU class 3 homeobox 1 OTF6 SCIP OTF-6 POU domain transcription factor SCIP POU domain class 3 transcription factor 1 octamer-binding protein 6 octamer-binding transcription factor 6
Gene Aliases: OCT6; OTF6; POU3F1; SCIP
UniProt ID: (Human) Q03052
Entrez Gene ID: (Human) 5453
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.