Invitrogen
Manufacturer Code:MA531460
Catalog # PIMA531460
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | GT735 |
Immunogen | Recombit protein encompassing a sequence within the center region of human Oct4. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: MGC22487 antibody OTF-3 antibody OTF3 antibody OTF4 antibody POU5F1 antibody POU-type homeodomain-containing DNA-binding protein antibody octamer-binding protein 3 antibody POU domain transcription factor OCT4 antibody octamer-binding protein 4 antibody POU domain class 5 transcription factor 1 antibody octamer-binding transcription factor 3 antibody octamer-binding transcription factor-3 antibody POU class 5 homeobox 1 antibody; Oct-3; Oct-4; Octamer-binding protein 3; Octamer-binding protein 4; Octamer-binding transcription factor 3; OTF-3; POU class 5 homeobox 1 transcript variant OCT4B1; POU class 5 homeobox 1 transcript variant OCT4B2; POU class 5 homeobox 1 transcript variant OCT4B3; POU class 5 homeobox 1 transcript variant OCT4B4; POU class 5 homeobox 1 transcript variant OCT4B5; POU class 5 homeobox 1 transcript variant OCT4B6; POU domain class 5 transcription factor 1; POU domain transcription factor OCT4; POU domain, class 5, transcription factor 1; POU-type homeodomain-containing DNA-binding protein
Gene Aliases: Oct-3; Oct-4; OCT3; OCT4; OTF-3; OTF3; OTF4; POU5F1
UniProt ID: (Human) Q01860
Entrez Gene ID: (Human) 5460
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.