Invitrogen
Manufacturer Code:PA572266
Catalog # PIPA572266
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Zebrafish |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 141-177 amino acids from the Central region of Zebrafish pou5f1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: POU domain class 5 transcription factor 1 POU domain protein 2 pou5f1 gp-9 pou-2 pou2; POU domain gene 2; POU domain protein 2; POU domain, class 5, transcription factor 1; pou-2; spiel-ohne-grenzen; spiel-ohne-grenzen/pou2
Gene Aliases: cb197; chunp6868; etID49452.21; gp-9; gp9; oct4; POU-2; pou2; pou5f1; spg; spg/pou2; wu:fd18d06
UniProt ID: (Zebrafish) Q90270
Entrez Gene ID: (Zebrafish) 30333
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.