Proteintech
Manufacturer Code:176951AP150UL
Catalog # 50-173-1726
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | OCRL Fusion Protein Ag11900 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Inositol polyphosphate 5-phosphatase OCRL; Inositol polyphosphate 5-phosphatase OCRL-1; INPP5F LOCR OCRL OCRL1; Lowe oculocerebrorenal syndrome protein; OCRL-1; oculocerebrorenal syndrome of Lowe; Phosphatidylinositol 3,4,5-triphosphate 5-phosphatase; phosphatidylinositol polyphosphate 5-phosphatase
Gene Aliases: 9530014D17Rik; BB143339; INPP5F; LOCR; NPHL2; OCRL; OCRL-1; OCRL1
UniProt ID: (Human) Q01968, (Mouse) Q6NVF0
Entrez Gene ID: (Human) 4952, (Rat) 317576, (Mouse) 320634
Molecular Function:
hydrolase
phosphatase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.