Invitrogen
Manufacturer Code:PA598335
Catalog # PIPA598335
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human P protein (2-136AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: eye color 2 (central brown); eye color 3 (brown); hair color 3 (brown); Melanocyte-specific transporter protein; oculocutaneous albinism II (pink-eye dilution homolog, mouse); P protein; P-protein; Pink-eyed dilution protein homolog; total brown iris pigmentation
Gene Aliases: BEY; BEY1; BEY2; BOCA; D15S12; EYCL; EYCL2; EYCL3; HCL3; OCA2; P; PED; SHEP1
UniProt ID: (Human) Q04671
Entrez Gene ID: (Human) 4948
Molecular Function:
cation transporter
transporter
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.